Ethyl 5-hydroxy-2-methylnicotinate 97%

Supplier: Ambeed
Warning

A289495-100MG A289495-250MG A289495-1G
103813-836EA 350 USD
103813-836 103834-976 76988-628 76988-630 77305-564
Ethyl 5-hydroxy-2-methylnicotinate 97%
Ethyl 5-hydroxy-2-methylnicotinate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18256626
CAS Number: 60390-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR