Ethyl-5-formyl-1H-pyrrole-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-formyl-1H-pyrrole-2-carboxylate

A211135-100MG A211135-250MG A211135-25G
103818-084EA 21 USD
103818-084 103848-776 77012-960 77012-962 77303-180
Ethyl-5-formyl-1H-pyrrole-2-carboxylate 97%
Ethyl-5-formyl-1H-pyrrole-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.1600 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00159462
CAS Number: 7126-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR