Ethyl-5-fluoro-1H-indole-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-fluoro-1H-indole-2-carboxylate, Ethyl 5-Fluoroindole-2-carboxylate, 5-Fluoroindole-2-carboxylic acid ethyl ester

A173724-5G A173724-1G
103806-864EA 18 USD
103806-864 103845-164 77082-434 77301-296
Ethyl-5-fluoro-1H-indole-2-carboxylate 98%
Ethyl-5-fluoro-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀FNO₂
MW: 207.21 g/mol
Boiling Pt: 312 °C (760 mm Hg)
Melting Pt: 146…150 °C
Flash Pt: 142.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00152076
CAS Number: 348-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR