Ethyl 5-chlorothiazolo[5,4-b]pyridine-2-carboxylate 95%

Supplier: Ambeed
Warning

A148099-100MG A148099-250MG A148099-1G
103803-878EA 861 USD
103803-878 77137-654 77137-656 77299-638
Ethyl 5-chlorothiazolo[5,4-b]pyridine-2-carboxylate 95%
Ethyl 5-chlorothiazolo[5,4-b]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇ClN₂O₂S
Storage Temperature: Refrigerator
MDL Number: MFCD26400831
CAS Number: 1202075-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR