Ethyl 5-chloropyrazolo[1,5-a]pyrimidine-3-carboxylate 97%

Supplier: Ambeed
Warning

A114913-100MG A114913-10G A114913-250MG A114913-1G A114913-5G A114913-25G A114913-100G
103798-500EA 9 USD
103798-500 103824-342 76869-950 76869-952 76869-954 77311-160 77311-162 77610-926 77670-610
Ethyl 5-chloropyrazolo[1,5-a]pyrimidine-3-carboxylate 97%
Ethyl 5-chloropyrazolo[1,5-a]pyrimidine-3-carboxylate

Precaución: for Research Use Only

Formula: C₉H₈ClN₃O₂
MW: 225.632 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12407819
CAS Number: 1224944-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR