Ethyl-5-chloro-4-methylisoxazole-3-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-chloro-4-methylisoxazole-3-carboxylate

A207438-1G A207438-5G A207438-250MG
103817-794EA 91 USD
103817-794 103848-476 77086-750 77086-752 77112-346
Ethyl-5-chloro-4-methylisoxazole-3-carboxylate 98%
Ethyl-5-chloro-4-methylisoxazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈ClNO₃
MW: 189.6 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11520431
CAS Number: 3356-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR