Ethyl-5-chloro-1,3,4-thiadiazole-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-chloro-1,3,4-thiadiazole-2-carboxylate

A189756-250MG A189756-1G
103816-220EA 39 USD
103816-220 103846-816 76799-956 77084-588
Ethyl-5-chloro-1,3,4-thiadiazole-2-carboxylate 97%
Ethyl-5-chloro-1,3,4-thiadiazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O₂S
MW: 192.6234 g/mol
Boiling Pt: 288.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11112114
CAS Number: 64837-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR