Ethyl 5-bromothiophene-3-carboxylate 95%

Supplier: Ambeed
Warning

A329543-5G A329543-10G
103825-528EA 15 USD
103825-528 103836-484 76839-452 76978-210
Ethyl 5-bromothiophene-3-carboxylate 95%
Ethyl 5-bromothiophene-3-carboxylate

Precaución: For research use only

Formula: C₇H₇BrO₂S
MW: 235.098 g/mol
Boiling Pt: 261 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD14155650
CAS Number: 170355-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR