Ethyl-5-bromopyrimidine-2-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-bromopyrimidine-2-carboxylate

A190706-250MG A190706-1G A190706-5G
103816-322EA 16 USD
103816-322 103846-924 76784-898 77084-740 77084-742
Ethyl-5-bromopyrimidine-2-carboxylate 95%
Ethyl-5-bromopyrimidine-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O₂
MW: 231.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12964050
CAS Number: 1197193-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR