Ethyl 5-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate 97%

Supplier: Ambeed
Warning

A300050-100MG A300050-250MG A300050-1G A300050-5G
103814-256EA 145 USD
103814-256 103835-406 77166-946 77166-948 77305-786 77305-788
Ethyl 5-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate 97%
Ethyl 5-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate

Precaución: for Research Use Only

Formula: C₉H₈BrN₃O₂
MW: 270.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26518651
CAS Number: 1436686-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR