Ethyl 5-bromo-6-hydroxypicolinate 98%

Supplier: Ambeed
Warning

A123652-100MG A123652-250MG A123652-1G A123652-5G
103800-422EA 207 USD
103800-422 76822-554 76822-556 77297-950 77297-952
Ethyl 5-bromo-6-hydroxypicolinate 98%
Ethyl 5-bromo-6-hydroxypicolinate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₃
MW: 246.058 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698334
CAS Number: 1214346-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR