Ethyl-5-bromo-2-methylnicotinate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 5-bromo-2-methylnicotinate
A178590-250MG
A178590-1G
A178590-25G
A178590-100G
A178590-10G
103807-358EA
20
USD
103807-358
103845-694
77029-338
77083-130
77083-132
77550-148
77567-630
Ethyl-5-bromo-2-methylnicotinate 97%
Ethyl-5-bromo-2-methylnicotinate
Precaución: For research use only.
Formula:
C₉H₁₀BrNO₂ Boiling Pt: 263 °C (760 mm Hg) Flash Pt: 112.88 °C Storage Temperature: Ambient |
MDL Number:
MFCD10698571 CAS Number: 129477-21-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...