Ethyl 5-bromo-1H-pyrazole-3-carboxylate 98%

Supplier: Ambeed
Warning

A105153-250MG A105153-1G A105153-100MG
103796-846EA 83 USD
103796-846 103822-574 76809-538 77309-448 77882-898
Ethyl 5-bromo-1H-pyrazole-3-carboxylate 98%
Ethyl 5-bromo-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O₂
Boiling Pt: 335.4±22.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD20275526
CAS Number: 1392208-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR