Ethyl-5-bromo-1H-indole-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-bromoindole-2-carboxylate, 5-Bromoindole-2-carboxylic acid ethyl ester, Ethyl 5-Bromoindole-2-carboxylate, Ethyl 5-bromo-1H-indole-2-carboxylate

A155484-100G A155484-10G A155484-500G A155484-1G A155484-25G A155484-5G
103804-768EA 5 USD
103804-768 103842-966 77273-124 77273-126 77273-128 77300-122 77318-690 77318-692
Ethyl-5-bromo-1H-indole-2-carboxylate 97%
Ethyl-5-bromo-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrNO₂
MW: 268.11 g/mol
Melting Pt: 165 °C
Flash Pt: 192.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00022701
CAS Number: 16732-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR