Ethyl-5-bromo-1H-imidazole-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-bromo-1H-imidazole-2-carboxylate

A157598-100MG A157598-10G A157598-250MG A157598-1G A157598-25G A157598-5G
103805-018EA 29 USD
103805-018 103843-220 76812-524 76812-526 76812-528 77300-264 77300-266 77300-268
Ethyl-5-bromo-1H-imidazole-2-carboxylate 98%
Ethyl-5-bromo-1H-imidazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O₂
MW: 219.04 g/mol
Flash Pt: 154 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09842485
CAS Number: 944900-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR