Ethyl-5-bromo-1,3,4-thiadiazole-2-carboxylate 97%

Supplier: Ambeed
Warning

A293805-250MG A293805-1G A293805-10G A293805-25G A293805-100MG
103813-990EA 26 USD
103813-990 103835-130 76787-602 76837-752 77668-304 77668-306 77872-380
Ethyl-5-bromo-1,3,4-thiadiazole-2-carboxylate 97%
Ethyl-5-bromo-1,3,4-thiadiazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O₂S
Storage Temperature: Refrigerator
MDL Number: MFCD12165919
CAS Number: 1030613-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR