Ethyl-5-aminovalerate hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-aminopentanoate hydrochloride

A287472-1G A287472-5G A287472-10G A287472-250MG A287472-25G
103813-756EA 19 USD
103813-756 103834-898 76837-448 76837-450 76977-524 76977-526 76977-528 77881-792
Ethyl-5-aminovalerate hydrochloride 95%
Ethyl-5-aminovalerate hydrochloride

Precaución: For research use only

Formula: C₇H₁₆ClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD12546205
CAS Number: 29840-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR