Ethyl-5-aminopyrazine-2-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A175506-1G
A175506-10G
A175506-100MG
A175506-250MG
103807-052EA
95
USD
103807-052
103845-360
77082-704
77163-690
77892-159
77892-160
Ethyl-5-aminopyrazine-2-carboxylate 98%
Ethyl-5-aminopyrazine-2-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₉N₃O₂ Boiling Pt: 365.6 °C (760 mm Hg) Melting Pt: 172…173 °C Flash Pt: 174.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD08437665 CAS Number: 54013-06-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...