5-Aminoisoxazole-4-carboxylic acid ethyl ester 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 5-aminoisoxazole-4-carboxylate
A189204-250MG
A189204-1G
A189204-5G
103816-166EA
17
USD
103816-166
103846-760
76769-834
77084-522
77084-524
5-Aminoisoxazole-4-carboxylic acid ethyl ester 95%
5-Aminoisoxazole-4-carboxylic acid ethyl ester
Precaución: for Research Use Only
Formula:
C₆H₈N₂O₃ MW: 156.14 g/mol Boiling Pt: 288.3 °C (760 mm Hg) Melting Pt: 70…75 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00014093 CAS Number: 34859-64-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...