Ethyl-5-amino-1-methyl-1H-pyrazole-4-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate

A219718-25G A219718-5G A219718-100G A219718-10G A219718-500G
103809-550EA 227 USD
103809-550 103849-686 77088-242 77088-244 77112-690 77112-692 77112-694
Ethyl-5-amino-1-methyl-1H-pyrazole-4-carboxylate 97%
Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₁N₃O₂
MW: 169.18 g/mol
Boiling Pt: 300.5 °C (760 mm Hg)
Melting Pt: 101…102 °C
Flash Pt: 135.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00051652
CAS Number: 31037-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR