Ethyl-5-amino-1H-pyrazole-4-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-amino-1H-pyrazole-4-carboxylate

A352202-100G A352202-10G A352202-500G A352202-25G A352202-5G
103837-060EA 12 USD
103837-060 77154-996 77154-998 77155-000 77306-624 77306-626
Ethyl-5-amino-1H-pyrazole-4-carboxylate 97%
Ethyl-5-amino-1H-pyrazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₆H₉N₃O₂
MW: 155.1546 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00114562
CAS Number: 1260243-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR