Ethyl-5-amino-1H-pyrazole-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-amino-1H-pyrazole-3-carboxylate

A107942-100G A107942-10G A107942-250MG A107942-25G A107942-1G A107942-5G
103797-270EA 10 USD
103797-270 103823-044 76796-634 76796-636 76796-638 76796-640 77283-664 77283-666
Ethyl-5-amino-1H-pyrazole-3-carboxylate 97%
Ethyl-5-amino-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₉N₃O₂
MW: 155.1600 g/mol
Flash Pt: 190.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03468385
CAS Number: 105434-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR