Ethyl 5-amino-1-(2-bromophenyl)-1H-pyrazole-4-carboxylate 95%

Supplier: Ambeed
Warning

A373677-250MG A373677-1G
103826-538EA 62 USD
103826-538 103837-556 76817-546 76840-824
Ethyl 5-amino-1-(2-bromophenyl)-1H-pyrazole-4-carboxylate 95%
Ethyl 5-amino-1-(2-bromophenyl)-1H-pyrazole-4-carboxylate
Formula: C₁₂H₁₂BrN₃O₂
Boiling Pt: 433.456 °C (760 mm Hg)
Flash Pt: 215.947 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06637280
CAS Number: 1019009-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR