Ethyl-5-(trifluoromethyl)picolinate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-(trifluoromethyl)-2-pyridinecarboxylate

A882088-1G A882088-5G A882088-250MG
103841-312EA 48 USD
103841-312 76845-582 76845-584 77006-954
Ethyl-5-(trifluoromethyl)picolinate 97%
Ethyl 5-(trifluoromethyl)picolinate

Precaución: for Research Use Only

Formula: C₉H₈F₃NO₂
MW: 219.16 g/mol
Boiling Pt: 264.7 °C (760 mm Hg)
Melting Pt: 45…48 °C
Flash Pt: 113.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11100197
CAS Number: 128072-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR