Ethyl 5-(hydroxymethyl)isoxazole-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: 5-(Hydroxymethyl)isoxazole-3-carboxylic acid ethyl ester

A171141-100MG A171141-250MG A171141-25G A171141-1G A171141-5G
103806-598EA 39 USD
103806-598 103844-872 77011-574 77011-576 77011-578 77082-002 77082-004
Ethyl 5-(hydroxymethyl)isoxazole-3-carboxylate 95%
Ethyl 5-(hydroxymethyl)isoxazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉NO₄
MW: 171.151 g/mol
Boiling Pt: 338.8±32.0 °C (760 mm Hg)
Flash Pt: 158.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08273501
CAS Number: 123770-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR