Ethyl-5-chloromethyl-2-furancarboxylate 97%

Supplier: Ambeed
Danger

Synonyms: 5-Chloromethyl-2-furancarboxylic Acid Ethyl Ester, Ethyl 5-(chloromethyl)furan-2-carboxylate

A277285-1G A277285-25G A277285-10G
103813-346EA 10 USD
103813-346 103853-280 77093-176 77093-178 77626-430
Ethyl-5-chloromethyl-2-furancarboxylate 97%
Ethyl-5-chloromethyl-2-furancarboxylate

Precaución: for Research Use Only

Formula: C₈H₉ClO₃
MW: 188.61 g/mol
Boiling Pt: 282.4 °C (760 mm Hg)
Flash Pt: 124.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00173837
CAS Number: 2528-00-9
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR