Ethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinate 97%

Supplier: Ambeed
Warning

Synonyms: 6-(Ethoxycarbonyl)pyridine-3-boronic acid pinacol ester

A199228-10G A199228-250MG A199228-1G A199228-5G
103817-082EA 44 USD
103817-082 103847-706 76986-382 76986-384 77302-614 77302-616
Ethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinate 97%
Ethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinate

Precaución: for Research Use Only

Formula: C₁₄H₂₀BNO₄
MW: 277.12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12923436
CAS Number: 741709-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR