Ethyl pyrimidine-4-carboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 4-pyrimidinecarboxylate
A318634-1G
A318634-250MG
A318634-5G
103825-178EA
37
USD
103825-178
103836-112
76838-968
77114-704
77306-184
Ethyl pyrimidine-4-carboxylate 95%
Ethyl pyrimidine-4-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₈N₂O₂ MW: 152.15 g/mol Boiling Pt: 240.3 °C (760 mm Hg) Melting Pt: 36…40 °C Flash Pt: 99 °C Storage Temperature: Ambient |
MDL Number:
MFCD09832999 CAS Number: 62846-82-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...