Ethyl-4-pyrazolecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: ethyl 1H-pyrazole-4-carboxylate, Ethyl 4-pyrazolecarboxylate

A276564-100G A276564-10G A276564-500G A276564-1G A276564-25G A276564-5G
103813-310EA 5 USD
103813-310 103853-244 76772-242 76772-244 76772-246 77093-108 77093-110 77093-112
Ethyl-4-pyrazolecarboxylate 98%
Ethyl-4-pyrazolecarboxylate

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂
Boiling Pt: 307.8 °C (760 mm Hg)
Melting Pt: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00010844
CAS Number: 37622-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR