Ethyl levulinate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-oxopentanoate, 4-Oxovaleric acid ethyl ester, Ethyl 4-oxovalerate, 4-Ketovaleric acid ethyl ester, 4-Oxopentanoic acid ethyl ester, Levulinic acid ethyl ester

A132069-100G A132069-1G A132069-500G A132069-5G A132069-1000G A132069-25G
103808-436EA 57 USD
103808-436 76972-124 76972-126 76972-128 76972-130 77298-532 77314-240
Ethyl levulinate 98%
Ethyl levulinate

Precaución: for Research Use Only

Formula: C₇H₁₂O₃
Boiling Pt: 205.5 °C (760 mm Hg)
Flash Pt: 77.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00009209
CAS Number: 539-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR