Ethyl-4-nitropyrrole-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 4-Nitropyrrole-2-carboxylic Acid Ethyl Ester, Ethyl 4-nitro-1H-pyrrole-2-carboxylate

A217700-25G A217700-10G A217700-250MG A217700-1G A217700-5G
103818-636EA 24 USD
103818-636 103849-366 77087-992 77165-134 77165-136 77303-458 77303-460
Ethyl-4-nitropyrrole-2-carboxylate 97%
Ethyl-4-nitropyrrole-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈N₂O₄
MW: 184.15 g/mol
Boiling Pt: 347.7 °C (760 mm Hg)
Melting Pt: 174 °C
Flash Pt: 164.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00059927
CAS Number: 5930-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR