Ethyl-4-methylthiazole-5-carboxylate 98%

Supplier: Ambeed
Warning

A193444-25G A193444-5G A193444-1G A193444-100G
103816-538EA 17 USD
103816-538 103847-154 76999-058 77085-058 77302-344 77881-431
Ethyl-4-methylthiazole-5-carboxylate 98%
Ethyl-4-methylthiazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉NO₂S
Boiling Pt: 239.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00626873
CAS Number: 20582-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR