Ethyl 4-methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)benzoate 97%

Supplier: Ambeed
Warning

A230580-10G A230580-1G A230580-25G A230580-5G
103819-556EA 7 USD
103819-556 103850-456 77031-034 77089-316 77089-318 77089-320
Ethyl 4-methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)benzoate 97%
Ethyl 4-methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)benzoate

Precaución: for Research Use Only

Formula: C₁₉H₁₈N₄O₂
MW: 334.37 g/mol
Boiling Pt: 546.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11100712
CAS Number: 641569-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR