Ethyl-4-methylpyrrole-3-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 4-methyl-1H-pyrrole-3-carboxylate
A261744-1G
A261744-5G
103812-532EA
15
USD
103812-532
103852-422
77304-820
77304-822
Ethyl-4-methylpyrrole-3-carboxylate 97%
Ethyl 4-methylpyrrole-3-carboxylate
Precaución: for Research Use Only
Formula:
C₈H₁₁NO₂ MW: 153.1800 g/mol Boiling Pt: 289.7 °C (760 mm Hg) Melting Pt: 72…75 °C Flash Pt: 129 °C Storage Temperature: Ambient |
MDL Number:
MFCD00030417 CAS Number: 2199-49-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...