Ethyl-4-iodobutyrate 96%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-iodobutanoate

A174381-10G A174381-1G A174381-25G A174381-5G A174381-250MG
103806-940EA 35 USD
103806-940 103845-240 76824-618 77082-542 77301-332 77301-334 77683-402
Ethyl-4-iodobutyrate 96%
Ethyl 4-iodobutyrate

Precaución: For research use only.

Formula: C₆H₁₁IO₂
MW: 242.0600 g/mol
Boiling Pt: 219.6 °C (760 mm Hg)
Flash Pt: 86.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00045325
CAS Number: 7425-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR