Ethyl-4-hydroxyquinoline-3-carboxylate 98%

Supplier: Ambeed
Warning

A219655-25G A219655-5G
103818-804EA 9 USD
103818-804 103849-682 77088-232 77088-234
Ethyl-4-hydroxyquinoline-3-carboxylate 98%
Ethyl 4-hydroxyquinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO₃
Boiling Pt: 343.7 °C (760 mm Hg)
Melting Pt: 280…281 °C
Storage Temperature: Ambient
MDL Number: MFCD00173406
CAS Number: 26892-90-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR