Ethyl-4-hydroxycyclohexanecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-hydroxycyclohexanecarboxylate

A280791-100G A280791-10G A280791-500G A280791-1G A280791-25G A280791-5G
103813-522EA 9 USD
103813-522 103853-470 76802-166 76802-168 76802-170 76837-158 76837-160 76837-162
Ethyl-4-hydroxycyclohexanecarboxylate 97%
Ethyl-4-hydroxycyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₉H₁₆O₃
Boiling Pt: 251.4 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00192156
CAS Number: 17159-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR