Ethyl-4-hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylate

A101662-10G A101662-25G A101662-1G A101662-5G
103796-074EA 15 USD
103796-074 103821-770 76717-560 76717-562 77308-692 77308-694
Ethyl-4-hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylate 95%
Ethyl 4-hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₃NO₄
MW: 301.2200 g/mol
Boiling Pt: 357.3 °C (760 mm Hg)
Flash Pt: 169.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00219847
CAS Number: 175203-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR