Ethyl-4H-thieno[3,2-b]pyrrole-5-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4H-thieno[3,2-b]pyrrole-5-carboxylate

A563619-1G A563619-25G A563619-5G A563619-10G A563619-250MG
103828-762EA 14 USD
103828-762 103839-898 76844-006 76844-008 76844-010 76980-276 76980-278
Ethyl-4H-thieno[3,2-b]pyrrole-5-carboxylate 98%
Ethyl-4H-thieno[3,2-b]pyrrole-5-carboxylate

Precaución: For research use only

Formula: C₉H₉NO₂S
MW: 195.2383 g/mol
Boiling Pt: 345.7 °C (760 mm Hg)
Melting Pt: 131…132 °C
Flash Pt: 162.9 °C
Storage Temperature: Ambient
MDL Number: MFCD03425800
CAS Number: 46193-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR