Ethyl 4-fluoropiperidine-4-carboxylate hydrochloride 97%

Supplier: Ambeed
Warning

A124864-100MG A124864-250MG A124864-5G A124864-1G
103800-584EA 41 USD
103800-584 76996-050 76996-052 76996-054 77312-726
Ethyl 4-fluoropiperidine-4-carboxylate hydrochloride 97%
Ethyl 4-fluoropiperidine-4-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₄FNO₂·ClH
MW: 211.662 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09880187
CAS Number: 845909-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR