Ethyl-4-fluoro-3-nitrobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-fluoro-3-nitrobenzoate

A349591-100G A349591-10G A349591-1G A349591-25G A349591-5G
103826-020EA 5 USD
103826-020 103837-012 76694-088 76694-090 76840-104 76840-106 76840-108
Ethyl-4-fluoro-3-nitrobenzoate 98%
Ethyl-4-fluoro-3-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₈FNO₄
MW: 213.1600 g/mol
Boiling Pt: 312.3 °C (760 mm Hg)
Melting Pt: 40…44 °C
Storage Temperature: Ambient
MDL Number: MFCD03428515
CAS Number: 367-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR