Ethyl 4-cyclopropyl-3-oxobutanoate 95%

Supplier: Ambeed
Warning

A225422-1G A225422-5G A225422-100MG A225422-250MG
103819-180EA 148 USD
103819-180 103850-078 77088-776 77088-778 77165-350 77165-352
Ethyl 4-cyclopropyl-3-oxobutanoate 95%
Ethyl 4-cyclopropyl-3-oxobutanoate

Precaución: for Research Use Only

Formula: C₉H₁₄O₃
Boiling Pt: 232.0±13.0 °C (760 mm Hg)
Flash Pt: 94.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06656674
CAS Number: 630399-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR