Ethyl 4-chloropicolinate 97%

Supplier: Ambeed
Warning

A152362-10G A152362-1G
103804-380EA 14 USD
103804-380 103842-548 76823-864 77299-944
Ethyl 4-chloropicolinate 97%
Ethyl 4-chloropicolinate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MW: 185.608 g/mol
Boiling Pt: 278.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09702465
CAS Number: 64064-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR