Ethyl-4-chloro-6-methyl-2-(methylthio)pyrimidine-5-carboxylate 97%

Supplier: Ambeed
Warning

A188730-250MG A188730-1G A188730-100G A188730-10G
103816-126EA 36 USD
103816-126 103846-718 76974-774 77302-144 77567-758 77881-407
Ethyl-4-chloro-6-methyl-2-(methylthio)pyrimidine-5-carboxylate 97%
Ethyl-4-chloro-6-methyl-2-(methylthio)pyrimidine-5-carboxylate

Precaución: For research use only.

Formula: C₉H₁₁ClN₂O₂S
Boiling Pt: 346.7±37.0 °C (760 mm Hg)
Flash Pt: 163.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08063161
CAS Number: 583878-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR