Ethyl-4-chloro-3-nitrobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-chloro-3-nitrobenzoate, 4-Chlorobenzoic acid ethyl ester

A271407-25G A271407-100G A271407-10G
103852-956EA 14 USD
103852-956 77092-734 77262-466 77262-468
Ethyl-4-chloro-3-nitrobenzoate 98%
Ethyl-4-chloro-3-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₈ClNO₄
MW: 229.62 g/mol
Boiling Pt: 326.2 °C (760 mm Hg)
Melting Pt: 57…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00219621
CAS Number: 16588-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR