Ethyl 4-chloro-2-cyclopropylpyrimidine-5-carboxylate 97%

Supplier: Ambeed
Warning

A173183-1G A173183-100MG A173183-250MG
103806-800EA 77 USD
103806-800 103845-094 77082-332 77259-996 77259-998
Ethyl 4-chloro-2-cyclopropylpyrimidine-5-carboxylate 97%
Ethyl 4-chloro-2-cyclopropylpyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClN₂O₂
MW: 226.65 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11047088
CAS Number: 1044770-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR