Ethyl-4-bromopicolinate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-bromopicolinate

A104162-250MG A104162-1G A104162-5G
103796-664EA 13 USD
103796-664 103822-378 76780-674 77309-242 77309-244
Ethyl-4-bromopicolinate 97%
Ethyl 4-bromopicolinate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MW: 230.0600 g/mol
Boiling Pt: 299.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09839150
CAS Number: 62150-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR