Ethyl-4-bromobutyrate 98%

Supplier: Ambeed
Danger

Synonyms: Ethyl 4-bromobutyrate

A133511-100G A133511-10G A133511-250MG A133511-500G A133511-5G A133511-1000G A133511-25G A133511-1G
103808-464EA 65 USD
103808-464 76996-524 76996-526 76996-528 76996-530 76996-532 77314-540 77314-542 77670-900
Ethyl-4-bromobutyrate 98%
Ethyl-4-bromobutyrate

Precaución: for Research Use Only

Formula: C₆H₁₁BrO₂
Boiling Pt: 198.3 °C (760 mm Hg)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00000259
CAS Number: 2969-81-5
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR