Ethyl 4-bromo-6-chloronicotinate 97%

Supplier: Ambeed
Warning

A326270-100MG A326270-250MG A326270-1G
103825-424EA 353 USD
103825-424 103836-372 76648-284 76648-286 77306-334
Ethyl 4-bromo-6-chloronicotinate 97%
Ethyl 4-bromo-6-chloronicotinate

Precaución: for Research Use Only

Formula: C₈H₇BrClNO₂
MW: 264.5 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26520840
CAS Number: 1807221-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR