Ethyl 4-bromo-5-methyl-1H-pyrazole-3-carboxylate 95%

Supplier: Ambeed
Warning

A100583-250MG A100583-1G A100583-5G
103795-524EA 102 USD
103795-524 103821-212 76821-210 77296-764 77296-766
Ethyl 4-bromo-5-methyl-1H-pyrazole-3-carboxylate 95%
Ethyl 4-bromo-5-methyl-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉BrN₂O₂
MW: 233.063 g/mol
Boiling Pt: 343.0±37.0 °C (760 mm Hg)
Melting Pt: 103…104 °C
Flash Pt: 161.2 °C
Storage Temperature: Ambient
MDL Number: MFCD11054303
CAS Number: 6076-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR